Learn More
Biolegend Purified anti-human CD38
Supplier:  Biolegend 356602
Encompass_Preferred
Purified anti-human CD38 [HB-7]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, ICC, WB; Size: 100 ug
Catalog No. 50-605-328