Learn More
Biolegend Purified anti-human MIF
Supplier:  Biolegend 525501
Encompass_Preferred
Purified anti-human MIF [10C3]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ELISA Capture, WB; Size: 50 ug
Catalog No. 50-208-5952