Learn More
Biolegend Purified anti-human/rat CD51 (Integrin αvβ5)
Supplier:  Biolegend 920004
Encompass_Preferred
Purified anti-human/rat CD51 (Integrin αvβ5) [P1F6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Rat, Cross-Reactivity: Hamster; Apps: FC; Size: 100 ug
Catalog No. 50-207-5228