Learn More
Biolegend Purified anti-human CD228 (MFI2, MTF1)
Supplier:  Biolegend 363101
Encompass_Preferred
Purified anti-human CD228 (MFI2, MTF1) [SHM72]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-207-8130