Learn More
Biolegend Purified anti-human CD68
Supplier:  Biolegend 375602
Encompass_Preferred
Purified anti-human CD68 [BL13756]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: IHC-P; Size: 100 ug
Catalog No. 50-207-8843