Learn More
Biolegend Purified anti-human CD99
Supplier:  Biolegend 398202
Encompass_Preferred
Purified anti-human CD99 [hec2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, Block, IHC, IP, WB; Size: 100 ug
Catalog No. 50-207-9350