Learn More
Biolegend Purified anti-human IgG2
Supplier:  Biolegend 411102
Encompass_Preferred
Purified anti-human IgG2 [HP6002]; Isotype: Mouse IgG1; Reactivity: Human; Apps: ELISA Capture; Size: 500 ug
Catalog No. 50-207-9897