Learn More
Biolegend Purified anti-HYOU1
Supplier:  Biolegend 671402
Encompass_Preferred
Purified anti-HYOU1 [W18144A]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: WB, IP, ICC; Size: 100 ug
Catalog No. 50-208-7299