Learn More
Biolegend Purified anti-IKKγ (NEMO)
Supplier:  Biolegend 694101
Encompass_Preferred
Purified anti-IKKγ (NEMO) [W16070A]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: WB, ICC; Size: 25 ug
Catalog No. 50-208-7647