Learn More
Biolegend Purified anti-IL-18
Supplier:  Biolegend 934802
Encompass_Preferred
Purified anti-IL-18 [W18160A]; Isotype: Rat IgG2a, λ; Reactivity: Human; Apps: WB, IP, ICC; Size: 100 ug
Catalog No. 50-207-5324