Learn More
Biolegend Purified anti-IL-36α
Supplier:  Biolegend 676402
Encompass_Preferred
Purified anti-IL-36α [O91D11]; Isotype: Mouse IgG1; Reactivity: Human; Apps: WB; Size: 100 ug
Catalog No. 50-208-7357