Learn More
Biolegend Purified anti-ILK
Supplier:  Biolegend 694902
Encompass_Preferred
Purified anti-ILK [P83A9]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, KO/KD-WB, ICC; Size: 100 ug
Catalog No. 50-208-7658