Learn More
Biolegend Purified anti-Keratin 1
Supplier:  Biolegend 905203
Encompass_Preferred
Purified anti-Keratin 1 [Poly19052]; Isotype: Rabbit Polyclonal; Reactivity: Human, Rat, Cross-Reactivity: Primate; Apps: IHC-P, IHC-F, WB; Size: 25 ug
Catalog No. 50-207-5154