Learn More
Biolegend Purified anti-Lamin A
Supplier:  Biolegend 613501
Encompass_Preferred
Purified anti-Lamin A [Poly6135]; Isotype: Rabbit Polyclonal IgG; Reactivity: Mouse, Human; Apps: WB, ICC; Size: 50 uL
Catalog No. 50-170-658