Learn More
Biolegend Purified anti-LAT
Supplier:  Biolegend 623902
Encompass_Preferred
Purified anti-LAT [LAT1111]; Isotype: Mouse IgG1; Reactivity: Human; Apps: WB, ICFC, IP; Size: 100 ug
Catalog No. 50-170-757