Learn More
Biolegend Purified anti-Lck
Supplier:  Biolegend 628302
Encompass_Preferred
Purified anti-Lck [LCK-01]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, ICC, IP, FC; Size: 100 ug
Catalog No. 50-170-814