Learn More
Biolegend Purified anti-Lck Phospho (Tyr505)
Supplier:  Biolegend 699702
Encompass_Preferred
Purified anti-Lck Phospho (Tyr505) [A17013A]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 ug
Catalog No. 50-208-7768