Learn More
Biolegend Purified anti-Lck Phospho (Tyr505)
Supplier:  Biolegend 684301
Encompass_Preferred
Purified anti-Lck Phospho (Tyr505) [A17013B]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: WB; Size: 25 ug
Catalog No. 50-208-7489