Learn More
Biolegend Purified anti-MFF
Supplier:  Biolegend 857501
Encompass_Preferred
Purified anti-MFF [N382/14]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: WB, IHC-P; Size: 25 ug
Catalog No. 50-207-4797