Learn More
Biolegend Purified anti-MINA
Supplier:  Biolegend 683302
Encompass_Preferred
Purified anti-MINA [1H6B44]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: WB, ICC; Size: 100 ug
Catalog No. 50-208-7468