Learn More
Biolegend Purified anti-MMP1
Supplier:  Biolegend 634702
Encompass_Preferred
Purified anti-MMP1 [F15P3B6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IHC-P; Size: 100 ug
Catalog No. 50-170-898