Learn More
Biolegend Purified anti-MMP8
Supplier:  Biolegend 688002
Encompass_Preferred
Purified anti-MMP8 [1502022]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: WB, IHC-P; Size: 100 ug
Catalog No. 50-208-7557