Learn More
Biolegend Purified anti-mouse CD130 (gp130)
Supplier:  Biolegend 149402
Encompass_Preferred
Purified anti-mouse CD130 (gp130) [4H1B35]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-1859