Learn More
Biolegend Purified anti-mouse CD14
Supplier:  Biolegend 150101
Encompass_Preferred
Purified anti-mouse CD14 [M14-23]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-1912