Learn More
Biolegend Purified anti-mouse CD321
Supplier:  Biolegend 158502
Encompass_Preferred
Purified anti-mouse CD321 [90G4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-207-2398