Learn More
Biolegend Purified anti-mouse Aiolos
Supplier:  Biolegend 653201
Encompass_Preferred
Purified anti-mouse Aiolos [8B2]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: WB, ICFC, IP; Size: 25 ug
Catalog No. 50-208-7031