Learn More
Biolegend Purified anti-mouse CD102
Supplier:  Biolegend 105602
Encompass_Preferred
Purified anti-mouse CD102 [3C4 (MIC2/4)]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F, IP; Size: 500 ug
Catalog No. 50-163-060