Learn More
Biolegend Purified anti-mouse CD115 (CSF-1R)
Supplier:  Biolegend 135501
Encompass_Preferred
Purified anti-mouse CD115 (CSF-1R) [AFS98]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, Block, IHC-F; Size: 50 ug
Catalog No. 50-164-846