Learn More
Biolegend Purified anti-mouse/human CD11b
Supplier:  Biolegend 101201
Encompass_Preferred
Purified anti-mouse/human CD11b [M1/70]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Chimpanzee,Rabbit (Lapine); Apps: FC, CyTOF, IP, IHC-F, ICC; Size: 50 ug
Catalog No. 50-162-520
Edge