Learn More
Biolegend Purified anti-mouse CD11c (Maxpar Ready)
Supplier:  Biolegend 117341
Encompass_Preferred
Purified anti-mouse CD11c (Maxpar Ready) [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, CyTOF; Size: 100 ug
Catalog No. 50-207-0292