Learn More
Biolegend Purified anti-mouse CD11c
Supplier:  Biolegend 117301
Encompass_Preferred
Purified anti-mouse CD11c [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, CyTOF, IP, IHC; Size: 50 ug
Catalog No. 50-163-700