Learn More
Biolegend Purified anti-mouse CD160
Supplier:  Biolegend 143002
Encompass_Preferred
Purified anti-mouse CD160 [7H1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-1417