Learn More
Biolegend Purified anti-mouse CD18
Supplier:  Biolegend 101402
Encompass_Preferred
Purified anti-mouse CD18 [M18/2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC, IP, WB; Size: 500 ug
Catalog No. 50-162-558