Learn More
Biolegend Purified anti-mouse CD19
Supplier:  Biolegend 115501
Encompass_Preferred
Purified anti-mouse CD19 [6D5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, CyTOF, IHC-F; Size: 50 ug
Catalog No. 50-163-538