Learn More
Biolegend Purified anti-mouse CD200R3
Supplier:  Biolegend 142302
Encompass_Preferred
Purified anti-mouse CD200R3 [Ba160]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, Depletion; Size: 200 ug
Catalog No. 50-165-255