Learn More
Biolegend Purified anti-mouse CD23
Supplier:  Biolegend 101602
Encompass_Preferred
Purified anti-mouse CD23 [B3B4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IP, Block, IHC-F; Size: 500 ug
Catalog No. 50-162-570