Learn More
Biolegend Purified anti-mouse CD24
Supplier:  Biolegend 101802
Encompass_Preferred
Purified anti-mouse CD24 [M1/69]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, WB, IHC-F, IHC-P; Size: 500 ug
Catalog No. 50-162-585