Learn More
Biolegend Purified anti-mouse CD25
Supplier:  Biolegend 102002
Encompass_Preferred
Purified anti-mouse CD25 [PC61]; Isotype: Rat IgG1, λ; Reactivity: Mouse; Apps: FC, IHC-F, IP, Block, Depletion; Size: 500 ug
Catalog No. 50-162-614