Learn More
Biolegend Purified anti-mouse CD254 (TRANCE, RANKL)
Supplier:  Biolegend 510002
Encompass_Preferred
Purified anti-mouse CD254 (TRANCE, RANKL) [IK22/5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IP, WB; Size: 500 ug
Catalog No. 50-170-072