Learn More
Biolegend Purified anti-mouse CD270 (HVEM)
Supplier:  Biolegend 136302
Encompass_Preferred
Purified anti-mouse CD270 (HVEM) [HMHV-1B18]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-164-902