Learn More
Biolegend Purified anti-mouse CD28
Supplier:  Biolegend 102102
Encompass_Preferred
Purified anti-mouse CD28 [37.51]; Isotype: Syrian Hamster IgG; Reactivity: Mouse; Apps: FC, IP, IHC-F, Costim, Block; Size: 500 ug
Catalog No. 50-162-637