Learn More
Biolegend Purified anti-mouse TLR4 (CD284)/MD2 Complex
Supplier:  Biolegend 117601
Encompass_Preferred
Purified anti-mouse TLR4 (CD284)/MD2 Complex [MTS510]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-P, WB, ICFC, IP; Size: 50 ug
Catalog No. 50-163-729