Learn More
Biolegend Purified anti-mouse CD3ε
Supplier:  Biolegend 155602
Encompass_Preferred
Purified anti-mouse CD3ε [KT3.1.1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 500 ug
Catalog No. 50-207-2219