Learn More
Biolegend Purified anti-mouse CD31
Supplier:  Biolegend 102501
Encompass_Preferred
Purified anti-mouse CD31 [MEC13.3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IP, Block, IHC-F; Size: 50 ug
Catalog No. 50-162-694