Learn More
Biolegend Purified anti-mouse CD317 (BST2, PDCA-1)
Supplier:  Biolegend 127102
Encompass_Preferred
Purified anti-mouse CD317 (BST2, PDCA-1) [129c1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IP; Size: 100 ug
Catalog No. 50-164-339