Learn More
Biolegend Purified anti-mouse CD326 (Ep-CAM) (Maxpar Ready)
Supplier:  Biolegend 118223
Encompass_Preferred
Purified anti-mouse CD326 (Ep-CAM) (Maxpar Ready) [G8.8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, CyTOF; Size: 100 ug
Catalog No. 50-850-326