Learn More
Biolegend Purified anti-mouse CD39
Supplier:  Biolegend 135702
Encompass_Preferred
Purified anti-mouse CD39 [5F2]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC, WB, ELISA; Size: 100 ug
Catalog No. 50-164-859