Learn More
Biolegend Purified anti-mouse CD40
Supplier:  Biolegend 102902
Encompass_Preferred
Purified anti-mouse CD40 [HM40-3]; Isotype: Armenian Hamster IgM; Reactivity: Mouse; Apps: FC, Block, Costim, IHC-F; Size: 500 ug
Catalog No. 50-162-736