Learn More
Biolegend Purified anti-mouse CD45
Supplier:  Biolegend 103101
Encompass_Preferred
Purified anti-mouse CD45 [30-F11]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC-F, CyTOF, IP, CMCD, IHC, WB; Size: 50 ug
Catalog No. 50-162-774