Learn More
Biolegend Purified anti-mouse CD47
Supplier:  Biolegend 127501
Encompass_Preferred
Purified anti-mouse CD47 [miap301]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 50 ug
Catalog No. 50-164-374